Novus Biologicals
Manufacturer Code:NBP273179UV
Catalog # NBP273179UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E8 |
Immunogen | Full length recombinant protein of human p120-catenin (NP_001078927) produced in HEK293T cells. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cadherin-associated Src substrate; Cadherin-associated Src substrate CAS catenin (cadherin-associated protein) delta 1 catenin delta-1 CTNND KIAA0384P120CAS p120 p120 catenin p120(cas) p120(CTN) p120cas P120CTN; CAS; catenin (cadherin-associated protein), delta 1; Catenin delta-1; p120 catenin; p120(cas); p120(ctn)
Gene Aliases: CAS; CTNND; CTNND1; KIAA0384; p120; p120(CAS); p120(CTN); P120CAS; P120CTN
UniProt ID: (Human) O60716
Entrez Gene ID: (Human) 1500
Molecular Function: cytoskeletal protein intermediate filament intermediate filament binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.