Abnova
Manufacturer Code:H00079991D01P
Catalog # 89-018-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OBFC1 (NP_079204.1 1 a.a. ~ 368 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human OBFC1 protein.
For Research Use Only
Protein Aliases: AAF-44; alpha accessory factor 44; bA541N10.2; CST complex subunit STN1; FLJ22559; oligonucleotide/oligosaccharide-binding fold containing 1; Oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; RP11-541N10.2; RPA-32; Suppressor of cdc thirteen homolog
Gene Aliases: AAF-44; AAF44; bA541N10.2; OBFC1; RPA-32; STN1
UniProt ID: (Human) Q9H668
Entrez Gene ID: (Human) 79991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.