Abnova
Manufacturer Code:H00004860A01
Catalog # 89-004-217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | NP (NP_000261 174 a.a. ~ 283 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant NP.
For Research Use Only
Protein Aliases: epididymis secretory sperm binding protein Li 156an; FLJ94043; FLJ97288; FLJ97312; HEL-S-156an; Inosine phosphorylase; Inosine-guanosine phosphorylase; MGC117396; MGC125915; MGC125916; PNP; PRO1837; PUNP; Purine nucleoside phosphorylase; purine-nucleoside:orthophosphate ribosyltransferase
Gene Aliases: NP; PNP; PRO1837; PUNP
UniProt ID: (Human) P00491
Entrez Gene ID: (Human) 4860
Molecular Function: phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.