Proteintech
Manufacturer Code:164681AP150UL
Catalog # 50-173-0965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | nucleobindin 1 Fusion Protein Ag9548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALNUC; CALNUC NUC NUCB1 nucleobindin 1; Nucleobindin-1
Gene Aliases: B230337F23Rik; C77483; CALNUC; MTEST82; NUC; Nucb; NUCB1
UniProt ID: (Human) Q02818, (Mouse) Q02819
Entrez Gene ID: (Human) 4924, (Mouse) 18220
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.