Abnova
Manufacturer Code:H00010002D01P
Catalog # 89-018-192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NR2E3 (NP_055064.1 1 a.a. ~ 410 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human NR2E3 protein.
For Research Use Only
Protein Aliases: ESCS; MGC49976; Nuclear receptor subfamily 2 group E member 3; nuclear receptor subfamily 2, group E, member 3; Photoreceptor-specific nuclear receptor; PNR; rd7; Retina-specific nuclear receptor; RNR; RP37
Gene Aliases: ESCS; NR2E3; PNR; rd7; RNR; RP37
UniProt ID: (Human) Q9Y5X4
Entrez Gene ID: (Human) 10002
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.