Abnova
Manufacturer Code:H00009572M05
Catalog # 89-001-142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H6 |
Immunogen | NR1D1 (AAH56148 1 a.a. ~ 614 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant NR1D1.
For Research Use Only
Protein Aliases: EAR-1; EAR1; hRev; nuclear receptor Rev-ErbA-alpha; Nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; Rev-erbA-alpha; Rev-ErbAalpha; THRA1; THRAL; V-erbA-related protein 1
Gene Aliases: ear-1; EAR1; HREV; NR1D1; THRA1; THRAL
UniProt ID: (Human) P20393
Entrez Gene ID: (Human) 9572
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.