Invitrogen
Manufacturer Code:PA532170
Catalog # PIPA532170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 55 of nrv21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG9261-PA; CG9261-PC; CG9261-PD; CG9261-PE; CG9261-PF; CG9261-PG; ectatic; GH13134p; IP16413p; Na+/K+ ATPase; Na,K-ATPase beta-subunit; Na[+]/K[+] ATPase; nervana; nervana2; nirvana; nirvana2; nrv2-PA; nrv2-PC; nrv2-PD; nrv2-PE; nrv2-PF; nrv2-PG; Protein nervana 2; Sodium/potassium-dependent ATPase subunit beta-2; Sodium/potassium-transporting ATPase subunit beta-2
Gene Aliases: 42kDa; CG9261; Dmel\CG9261; Dmel_CG9261; ecta; K-ATPase; l(2)k04223; l(2)k13315; l(2)SH0329; l(2)SH2 0329; Na; Na[+]/K[+] ATPase; Na[+]/K[+]-ATPase; Na[+]/K[+]ATPase; NaK ATPase; NEST:bs13c10; Nrv; Nrv2; Nrv2.2
UniProt ID: (Fruit fly) Q24048
Entrez Gene ID: (Fruit fly) 33953
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.