Abnova
Manufacturer Code:H00004852M01
Catalog # 89-019-072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B5 |
Immunogen | NPY (AAH29497 29 a.a. ~ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant NPY.
For Research Use Only
Protein Aliases: C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y; NPY; prepro-neuropeptide Y; Pro-neuropeptide Y; PYY4
Gene Aliases: NPY; PYY4
UniProt ID: (Human) P01303
Entrez Gene ID: (Human) 4852
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.