Proteintech
Manufacturer Code:108881AP150UL
Catalog # 50-172-6686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | neuromedin B Fusion Protein Ag0842 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neuromedin B NMB; Neuromedin-B; Neuromedin-B-32; neuromedin-beta
Gene Aliases: 3110023K12Rik; NMB; RGD1562710
UniProt ID: (Human) P08949, (Mouse) Q9CR53
Entrez Gene ID: (Human) 4828, (Mouse) 68039, (Rat) 499194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.