Abnova
Manufacturer Code:H00004760M02
Catalog # 89-000-830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D11 |
Immunogen | NEUROD1 (AAH09046 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant NEUROD1.
For Research Use Only
Protein Aliases: basic helix-loop-helix transcription factor; beta-cell E-box transactivator 2; BETA2; BHF-1; bHLHa3; Class A basic helix-loop-helix protein 3; NEUROD; Neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD
Gene Aliases: BETA2; BHF-1; BHLHA3; MODY6; NEUROD; NEUROD1
UniProt ID: (Human) Q13562
Entrez Gene ID: (Human) 4760
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.