Abnova
Manufacturer Code:H00023327M04
Catalog # 89-001-225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2 |
Immunogen | NEDD4L (AAH32597 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant NEDD4L.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; hNedd4-2; KIAA0439; NEDD4-2; NEDD4.2; RSP5; ubiquitin-protein ligase Rsp5
Gene Aliases: hNEDD4-2; KIAA0439; NEDD4-2; NEDD4.2; NEDD4L; NEDL3; RSP5
UniProt ID: (Human) Q96PU5
Entrez Gene ID: (Human) 23327
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.