Novus Biologicals
Manufacturer Code:MAB2416G
Catalog # MAB2416G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 60718 |
Immunogen | Partial recombinant human nNOS protein (amino acids 218-1434) [UniProt P29475] |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bNOS; bNOS brain Constitutive NOS EC 1.14.13.39 IHPS1 Neuronal NOS nitric oxide synthase 1 (neuronal) N-NOS nNOSNOS type I NOS Peptidyl-cysteine S-nitrosylase NOS1; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxide synthase 1 (neuronal); Nitric oxide synthase, brain; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: bNOS; IHPS1; N-NOS; NC-NOS; nNOS; NOS; NOS1
UniProt ID: (Human) P29475
Entrez Gene ID: (Human) 4842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.