Invitrogen
Manufacturer Code:OSN00003W
Catalog # PIOSN00003W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic pentamer-peptide from the c-terminal region of human NOS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BNOS; BNOS NOS type I Neuronal NOS N-NOS nNOS Constitutive NOS NC-NOS Nitric oxide synthase; Constitutive NOS; N-NOS; NC-NOS; neuronal nitric oxide synthase; Neuronal NOS; nitric oxidase synthase; nitric oxide synthase 1 (neuronal); Nitric oxide synthase, brain; nNOS; nNOS; NOS type I; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: 2310005C01Rik; Bnos; IHPS1; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; NOS-I; NOS1
UniProt ID: (Human) P29475, (Rabbit) O19132, (Rat) P29476, (Mouse) Q9Z0J4
Entrez Gene ID: (Human) 4842, (Rabbit) 100009243, (Rat) 24598, (Mouse) 18125, (Guinea pig) 100729682
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.