Novus Biologicals
Manufacturer Code:NB1203511
Catalog # NB1203511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues T(724) K R R A I G F K K L A E A V K(739) C of rat bNOS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bNOS; bNOS brain Constitutive NOS EC 1.14.13.39 IHPS1 Neuronal NOS nitric oxide synthase 1 (neuronal) N-NOS nNOSNOS type I NOS Peptidyl-cysteine S-nitrosylase NOS1; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxide synthase 1 (neuronal); Nitric oxide synthase, brain; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: bNOS; IHPS1; N-NOS; NC-NOS; nNOS; NOS; NOS1
UniProt ID: (Human) P29475
Entrez Gene ID: (Human) 4842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.