R&D Systems
Manufacturer Code:AF2416SP
Catalog # AF2416SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human nNOS Ser218-Ser1434 Accession # P29475 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bNOS; bNOS brain Constitutive NOS EC 1.14.13.39 IHPS1 Neuronal NOS nitric oxide synthase 1 (neuronal) N-NOS nNOSNOS type I NOS NOS1 Peptidyl-cysteine S-nitrosylase NOS1; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxide synthase 1 (neuronal); Nitric oxide synthase, brain; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: bNOS; IHPS1; N-NOS; NC-NOS; nNOS; NOS; NOS1
UniProt ID: (Human) P29475
Entrez Gene ID: (Human) 4842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.