Invitrogen
Manufacturer Code:PA136152
Catalog # 01-672-105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic fragment of rat cerebellar NO-synthase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BNOS; Constitutive NOS; IHPS1 NOS nNOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxidase synthase; nitric oxide synthase 1 (neuronal); Nitric oxide synthase, brain; nNOS; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: Bnos; IHPS1; N-NOS; NC-NOS; nNOS; NOS; NOS1
UniProt ID: (Human) P29475, (Rat) P29476
Entrez Gene ID: (Human) 4842, (Rat) 24598
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.