Novus Biologicals
Manufacturer Code:NBP288927
Catalog # NB036516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL9025 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: DDAFKDSPNPSEHGHSDQR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°:C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ10639 FLJ21066 FLJ22268 KIAA0799FLJ43256 MGC131988 myosin X myosin-X Unconventional myosin-10; myosin-X; Unconventional myosin-10; Unconventional myosin-X; unconventionnal myosin-X
Gene Aliases: KIAA0799; MYO10
UniProt ID: (Human) Q9HD67
Entrez Gene ID: (Human) 4651
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.