R&D Systems
Manufacturer Code:MAB6866SP
Catalog # MAB6866SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 677014 |
Immunogen | Synthetic peptide corresponding to His385-Pro398 of rat &mu Opioid R/OPRM1 Accession # P33535 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hMOP KIAA0403 LMOR MOP mOpioidR MOR MOR1 M-OR-1 MOR-1 MOR1MOR Mu opiate receptor Mu opioid receptor mu-type opioid receptor opioid receptor mu 1 OPRM OPRM1; M-OR-1; MOP; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; Mu-type opioid receptor; opioid receptor, mu 1
Gene Aliases: LMOR; M-OR-1; MOP; MOR; MOR1; OPRM; OPRM1
UniProt ID: (Human) P35372
Entrez Gene ID: (Human) 4988
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.