Novus Biologicals
Manufacturer Code:NBP272825G
Catalog # NBP272825G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G7 |
Immunogen | Full length recombinant protein of human mu Crystallin (NP_001879) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: crystallin mu DFNA40NADP-regulated thyroid-hormone binding protein mu-crystallin homolog NADP-regulated thyroid-hormone-binding protein THBP; crystallin, mu; Ketimine reductase mu-crystallin; mu-crystallin homolog; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; thiomorpholine-carboxylate dehydrogenase
Gene Aliases: CRYM; DFNA40; THBP
UniProt ID: (Human) Q14894
Entrez Gene ID: (Human) 1428
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.