Abnova
Manufacturer Code:H00004216M08
Catalog # 89-000-803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F10 |
Immunogen | MAP3K4 (NP_005913 1201 a.a. ~ 1300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant MAP3K4.
For Research Use Only
Protein Aliases: dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); FLJ42439; KIAA0213; MAP three kinase 1; MAP/ERK kinase kinase 4; MAPK/ERK kinase kinase 4; MAPKKK4; MEK kinase 4; MEKK4; Mitogen-activated protein kinase kinase kinase 4; MTK1; predicted protein of HQ0412; PRO0412; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of
Gene Aliases: KIAA0213; MAP3K4; MAPKKK4; MEKK 4; MEKK4; MTK1; PRO0412
UniProt ID: (Human) Q9Y6R4
Entrez Gene ID: (Human) 4216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.