Abnova
Manufacturer Code:H00004141B01
Catalog # 89-005-733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MARS (NP_004981.2 1 a.a. ~ 900 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human MARS protein.
For Research Use Only
Protein Aliases: cytosolic methionyl-tRNA synthetase; FLJ35667; Methionine--tRNA ligase, cytoplasmic; Methionyl-tRNA synthetase; METRS; MTRNS
Gene Aliases: CMT2U; ILFS2; ILLD; MARS; MARS1; METRS; MRS; MTRNS; SPG70
UniProt ID: (Human) P56192
Entrez Gene ID: (Human) 4141
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.