Abnova
Manufacturer Code:H00004354B01
Catalog # 89-005-782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPP1 (NP_002427.1 1 a.a. ~ 466 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human MPP1 protein.
For Research Use Only
Protein Aliases: 55 kDa erythrocyte membrane protein; AAG12; aging-associated gene 12; DXS552; DXS552E; EMP55; erythrocyte membrane protein p55; Membrane protein, palmitoylated 1; membrane protein, palmitoylated 1, 55kDa; migration-related gene 1; MRG1; p55; palmitoylated erythrocyte membrane protein; PEMP
Gene Aliases: AAG12; DXS552E; EMP55; MPP1; MRG1; PEMP
UniProt ID: (Human) Q00013
Entrez Gene ID: (Human) 4354
Molecular Function: cell junction protein kinase nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.