Abnova
Manufacturer Code:H00005469M02
Catalog # 89-000-891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G3 |
Immunogen | PPARBP (NP_004765 1391 a.a. ~ 1490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PPARBP.
For Research Use Only
Protein Aliases: Activator-recruited cofactor 205 kDa component; ARC205; CRSP1; CRSP200; DRIP205; DRIP230; Mediator complex subunit 1; Mediator of RNA polymerase II transcription subunit 1; MGC71488; p53 regulatory protein RB18A; PBP; Peroxisome proliferator-activated receptor-binding protein; PPAR binding protein; PPAR-binding protein; PPARBP; PPARG binding protein; PPARGBP; RB18A; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; Thyroid receptor-interacting protein 2; TR-interacting protein 2; TRAP220; TRIP-2; TRIP2; vitamin D receptor-interacting protein 230 kD; Vitamin D receptor-interacting protein complex component DRIP205
Gene Aliases: ARC205; CRSP1; CRSP200; DRIP205; DRIP230; MED1; PBP; PPARBP; PPARGBP; RB18A; TRAP220; TRIP2
UniProt ID: (Human) Q15648
Entrez Gene ID: (Human) 5469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.