Abnova
Manufacturer Code:H00004199M03
Catalog # 89-000-797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H5 |
Immunogen | ME1 (AAH25246 1 a.a. ~ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ME1.
For Research Use Only
Protein Aliases: HUMNDME; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; MES; NADP-dependent malic enzyme; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: HUMNDME; ME1; MES
UniProt ID: (Human) P48163
Entrez Gene ID: (Human) 4199
Molecular Function: acyltransferase decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.