Invitrogen
Manufacturer Code:PA577430
Catalog # PIPA577430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ENFG(S)KLGSHGKR corresponding to amino acid residues 365- 377 of rat mGluR8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member H; G protein-coupled receptor family C group 1 member H; G protein-coupled receptor, family C, group 1, member H; glutamate receptor, metabotropic 8; Metabotropic glutamate receptor 8; Metabotropic Glutamate Receptor 8 GRM8 mGlu8; metabotropic glutamate receptor subtype 8b; mGluR8
Gene Aliases: A230002O04; GLUR8; GPRC1H; GRM8; mGlu8; mGlur; MGLUR8; mGluR8b
UniProt ID: (Human) O00222, (Rat) P70579, (Mouse) P47743
Entrez Gene ID: (Human) 2918, (Rat) 60590, (Mouse) 14823
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.