Novus Biologicals
Manufacturer Code:NBP261857
Catalog # NBP2-61857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C1C1 |
Immunogen | Purified recombinant fragment of human GRM8 (AA: extra 440-583) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ41058 glutamate metabotropic receptor 8 glutamate receptor metabotropic 8 GPRC1HMGLUR8GLUR8 metabotropic glutamate receptor 8 MGC126724 mGlu8 mGluR8; glutamate receptor, metabotropic 8; Metabotropic glutamate receptor 8; mGluR8
Gene Aliases: GLUR8; GPRC1H; GRM8; mGlu8; MGLUR8
UniProt ID: (Human) O00222
Entrez Gene ID: (Human) 2918
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.