R&D Systems
Manufacturer Code:MAB5277
Catalog # MAB5277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 476410 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human mGluR8 Gln34-Ser514 (predicted) Accession # O00222 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ41058 glutamate metabotropic receptor 8 glutamate receptor metabotropic 8 GPRC1H GPRC1HMGLUR8GLUR8 GRM8 metabotropic glutamate receptor 8 MGC126724 mGlu8 mGluR8; glutamate receptor, metabotropic 8; Metabotropic glutamate receptor 8; mGluR8
Gene Aliases: GLUR8; GPRC1H; GRM8; mGlu8; MGLUR8
UniProt ID: (Human) O00222
Entrez Gene ID: (Human) 2918
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.