Novus Biologicals
Manufacturer Code:NBP261856
Catalog # NBP2-61856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G8D3 |
Immunogen | Purified recombinant fragment of human GRM7 (AA: 454-590) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ40498 glutamate receptor metabotropic 7 metabotropic glutamate receptor 7 mGlu7 mGluR7 MGLUR7GPRC1GGLUR7; glutamate receptor, metabotropic 7; Metabotropic glutamate receptor 7; mGluR7; protein phosphatase 1, regulatory subunit 87
Gene Aliases: GLUR7; GPRC1G; GRM7; MGLU7; MGLUR7; PPP1R87
UniProt ID: (Human) Q14831
Entrez Gene ID: (Human) 2917
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.