R&D Systems
Manufacturer Code:FAB4405N100UG
Catalog # FAB4405N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 437310 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human mGluR7 Gln35-Ile915 Accession # Q14831 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ40498 glutamate receptor metabotropic 7 GPRC1G GRM7 metabotropic glutamate receptor 7 mGlu7 mGluR7 MGLUR7GPRC1GGLUR7; glutamate receptor, metabotropic 7; Metabotropic glutamate receptor 7; mGluR7; protein phosphatase 1, regulatory subunit 87
Gene Aliases: GLUR7; GPRC1G; GRM7; MGLU7; MGLUR7; PPP1R87
UniProt ID: (Human) Q14831
Entrez Gene ID: (Human) 2917
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.