Invitrogen
Manufacturer Code:PA533826
Catalog # PIPA533826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from 2nd extracellular domain of human GRM7 / MGLUR7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor, metabotropic 7; GRM7 GLUR7 MGLU7 MGLUR7 GPRC1G; Metabotropic glutamate receptor 7; mGluR7; protein phosphatase 1, regulatory subunit 87
Gene Aliases: GLUR7; GPRC1G; GRM7; MGLU7; MGLUR7; PPP1R87
UniProt ID: (Human) Q14831
Entrez Gene ID: (Human) 2917
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.