Invitrogen
Manufacturer Code:PA5115575
Catalog # PIPA5115575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human mGluR7 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6330570A01Rik BB176677 C030018L03 E130018M02Rik FLJ40498 G protein-coupled receptor GRM7 G protein-coupled receptor family C group 1 member G GLUR7 glutamate metabotropic receptor 7 glutamate receptor glutamate receptor metabotropic 7 Gpr1g Gprc1g Grm7 metabotropic glutamate receptor 7 metabotropic glutamate receptor mGluR7 metabotropic glutamate receptor subtype 7b MGLU7 mGlu7a receptor mGluR7 PPP1R87 protein phosphatase 1 regulatory subunit 87; G protein-coupled receptor GRM7; G protein-coupled receptor, family C, group 1, member G; glutamate receptor, metabotropic 7; Metabotropic glutamate receptor 7; metabotropic glutamate receptor mGluR7; metabotropic glutamate receptor subtype 7b; mGlu7a receptor; mGluR7; protein phosphatase 1, regulatory subunit 87
Gene Aliases: 6330570A01Rik; BB176677; C030018L03; E130018M02Rik; GLUR7; Gpr1g; GPRC1G; GRM7; MGLU7; MGLUR7; PPP1R87
UniProt ID: (Human) Q14831, (Rat) P35400, (Mouse) Q68ED2
Entrez Gene ID: (Human) 2917, (Rat) 81672, (Mouse) 108073
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.