Invitrogen
Manufacturer Code:PA577429
Catalog # PIPA577429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GQSDDSTRK(S)TGEE corresponding to amino acid residues 368-381 of rat mGluR6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member F; glutamate receptor, metabotropic 6; GRM6; Metabotropic glutamate receptor 6; mGluR6
Gene Aliases: BC021919; CSNB1B; Gm3; GPRC1F; GRM6; mGlu6; MGLUR6; nerg1; nob2; nob3; nob4
UniProt ID: (Human) O15303, (Rat) P35349, (Mouse) Q5NCH9
Entrez Gene ID: (Human) 2916, (Rat) 24419, (Mouse) 108072
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.