Novus Biologicals
Manufacturer Code:NB300189
Catalog # NB300189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminus of rat metabotropic Glutamate Receptor 6 (mGluR6) [Uniprot: P35349] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSNB1B glutamate receptor metabotropic 6 GPRC1Fmetabotropic glutamate receptor 6 mGlu6 mGluR6 MGLUR6DKFZp686H1993; glutamate receptor, metabotropic 6; Metabotropic glutamate receptor 6; mGluR6
Gene Aliases: CSNB1B; GPRC1F; GRM6; mGlu6; MGLUR6
UniProt ID: (Human) O15303
Entrez Gene ID: (Human) 2916
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.