Invitrogen
Manufacturer Code:PA5111826
Catalog # PIPA5111826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acids 367-380 of rat mGluR5 (Extracellular N-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: 6430542K11Rik A830007B11Rik AI850523 D16Ium24 D16Ium24e Glu5R Glur5 Glur-5 Glurbeta1 GPRC1A GPRC1E GRM 1 GRM 5 GRM1 GRM1 alpha GRM1A Grm5 metabotropic 1 Metabotropic glutamate receptor 5 MGC25101 mGlu1 mGlu5 mGluR mGluR1 mGluR1 alpha mGluR1A Mglur5 mGluR5a mGluR5b PPP1R86; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR5
Gene Aliases: 6430542K11Rik; AI850523; Glu5R; Gprc1e; Grm5; Mglur5; mGluR5b
UniProt ID: (Rat) P31424, (Mouse) Q3UVX5
Entrez Gene ID: (Rat) 24418, (Mouse) 108071
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.