Invitrogen
Manufacturer Code:MA527690
Catalog # PIMA527690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S75-33 |
Immunogen | Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; Glutamate receptor metabotropic 5GPRC1E GRM 5 GRM5 Metabotropic glutamate receptor 5 Metabotropic glutamate receptor 5 variant F Metabotropic glutamate receptor 5 variant G Metabotropic glutamate receptor 5 variant H mGlu5 mGluR5 mGluR5a mGluR5b Glutamate receptor metabotropic 1 glutamate receptor metabotropic 1 GPRC1A GRM 1 GRM1 alpha GRM1 GRM1&lowbarHUMAN GRM1A Metabotropic glutamate receptor 1 mGlu1 mGluR1 alpha mGluR1 mGluR1A; glutamate receptor, metabotropic 5; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: 6430542K11Rik; AI850523; Glu5R; GPRC1E; GRM5; mGlu5; MGLUR5; mGluR5b; PPP1R86
UniProt ID: (Human) P41594, (Rat) P31424, (Mouse) Q3UVX5
Entrez Gene ID: (Human) 2915, (Rat) 24418, (Mouse) 108071
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.