Invitrogen
Manufacturer Code:MA538097
Catalog # PIMA538097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0250 |
Immunogen | A synthesized peptide derived from human GRM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 6430542K11Rik A830007B11Rik AI850523 D16Ium24 D16Ium24e G protein coupled receptor family C group 1 member E G protein-coupled receptor GRM5 G protein-coupled receptor family C group 1 member E Glu5R Glur5 Glur-5 Glurbeta1 glutamate metabotropic receptor 5 glutamate receptor glutamate receptor metabotropic 5 GPRC1A Gprc1e GRM 1 GRM 5 GRM1 GRM1 alpha GRM1A Grm5 metabotropic 1 metabotropic glutamate receptor (mGluR5) metabotropic glutamate receptor 5 metabotropic glutamate receptor 5b metabotropic glutamate receptor subtype 5b metabotropic glutamate receptor type 5 MGC25101 mGlu1 mGlu5 mGluR mGluR1 mGluR1 alpha mGluR1A mGlur5 mGluR5a mGluR5b PPP1R86 protein phosphatase 1 regulatory subunit 86; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; glutamate receptor, metabotropic 5; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: 6430542K11Rik; AI850523; Glu5R; GPRC1E; GRM5; mGlu5; MGLUR5; mGluR5b; PPP1R86
UniProt ID: (Human) P41594, (Rat) P31424, (Mouse) Q3UVX5
Entrez Gene ID: (Human) 2915, (Rat) 24418, (Mouse) 108071
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.