R&D Systems
Manufacturer Code:MAB4514SP
Catalog # MAB4514SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 464818 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human mGluR5 Gln21-Ser509 Accession # P41594 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor metabotropic 5 GPRC1E GPRC1EMGLUR5metabotropic glutamate receptor 5 GRM5 mGlu5 mGluR5; glutamate receptor, metabotropic 5; Metabotropic glutamate receptor 5; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: GPRC1E; GRM5; mGlu5; MGLUR5; PPP1R86
UniProt ID: (Human) P41594
Entrez Gene ID: (Human) 2915
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.