Invitrogen
Manufacturer Code:PA581187
Catalog # PIPA581187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Human GRM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A830007B11Rik D16Ium24 D16Ium24e Glur-5 Glur5 Glurbeta1 MGC25101; glutamate receptor, metabotropic 5; Metabotropic glutamate receptor 5; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: GPRC1E; GRM5; mGlu5; MGLUR5; PPP1R86
UniProt ID: (Human) P41594
Entrez Gene ID: (Human) 2915
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.