Invitrogen
Manufacturer Code:PA532494
Catalog # PIPA532494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from C-terminus of human mGluR5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor, metabotropic 5; GPRC1E MGLUR5 mGlu5 metabotropic glutamate receptor 5 metabotropic glutamate receptor 5 variant F metabotropic glutamate receptor 5 variant G metabotropic glutamate receptor 5 variant H; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: GPRC1E; GRM5; mGlu5; MGLUR5; PPP1R86
UniProt ID: (Human) P41594, (Rat) P31424
Entrez Gene ID: (Human) 2915, (Rat) 24418
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.