Invitrogen
Manufacturer Code:PA513361
Catalog # PIPA513361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1020-1050 amino acids from the C-terminal region of human Metabotropic Glutamate Receptor 5 (GPRC1E) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; glutamate receptor, metabotropic 5; Metabotropic glutamate receptor 5; Metabotropic glutamate receptor 5 &lsqbPrecursor] mGluR5 GRM5 GPRC1E MGLUR5; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR5; protein phosphatase 1, regulatory subunit 86
Gene Aliases: 6430542K11Rik; AI850523; Glu5R; GPRC1E; GRM5; mGlu5; MGLUR5; mGluR5b; PPP1R86
UniProt ID: (Human) P41594, (Mouse) Q3UVX5
Entrez Gene ID: (Human) 2915, (Mouse) 108071
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.