Invitrogen
Manufacturer Code:720375
Catalog # PI720375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to Human GRM4 (aa 33-46 ) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutamate Metabotropic Receptor 4; glutamate receptor, metabotropic 4; Metabotropic glutamate receptor 4; mGluR4
Gene Aliases: GPRC1D; GRM4; mGlu4; MGLUR4
UniProt ID: (Human) Q14833
Entrez Gene ID: (Human) 2914
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.