Novus Biologicals
Manufacturer Code:NBP303682100UL
Catalog # NB092615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 33-290 of human mGluR4 (NP_000832.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor metabotropic 4 GPRC1Dmetabotropic glutamate receptor 4 MGC177594 mGlu4 mGluR4 MGLUR4glutamate metabotropic receptor 4; glutamate receptor, metabotropic 4; Metabotropic glutamate receptor 4; mGluR4
Gene Aliases: GPRC1D; GRM4; mGlu4; MGLUR4
UniProt ID: (Human) Q14833
Entrez Gene ID: (Human) 2914
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.