R&D Systems
Manufacturer Code:AF7387SP
Catalog # AF7387SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human mGluR4 Lys33-Ser518 Accession # Q14833 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor metabotropic 4 GPRC1D GPRC1Dmetabotropic glutamate receptor 4 GRM4 MGC177594 mGlu4 mGluR4 MGLUR4glutamate metabotropic receptor 4; glutamate receptor, metabotropic 4; Metabotropic glutamate receptor 4; mGluR4
Gene Aliases: GPRC1D; GRM4; mGlu4; MGLUR4
UniProt ID: (Human) Q14833
Entrez Gene ID: (Human) 2914
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.