Invitrogen
Manufacturer Code:PA577420
Catalog # PIPA577420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GDHNFMRREIKIEGD corresponding to amino acids 24-38 of rat mGluR3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor, family C, group 1, member C; GRM3; Metabotropic glutamate receptor 3; mGluR3
Gene Aliases: 0710001G23Rik; Gprc1c; Grm3; mGlu3; Mglur3
UniProt ID: (Mouse) Q9QYS2, (Rat) P31422
Entrez Gene ID: (Mouse) 108069, (Rat) 24416
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.