Novus Biologicals
Manufacturer Code:NBP261842
Catalog # NBP2-61842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6H10C3 |
Immunogen | Purified recombinant fragment of human GRM3 (AA: extra 433-576) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: GLUR3 glutamate receptor metabotropic 3 GPRC1Cglutamate metabotropic receptor 3 GRM3 mGlu3 mGluR3 MGLUR3metabotropic glutamate receptor 3; glutamate receptor, metabotropic 3; Metabotropic glutamate receptor 3; mGluR3
Gene Aliases: GLUR3; GPRC1C; GRM3; mGlu3; MGLUR3
UniProt ID: (Human) Q14832
Entrez Gene ID: (Human) 2913
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.