Invitrogen
Manufacturer Code:PA129424
Catalog # PIPA129424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide TKVDEAEYM corresponding to amino acids 118-126 of Rat metabotropic Glutamate Receptor 3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor, family C, group 1, member C; glutamate receptor, metabotropic 3; Metabotropic glutamate receptor 3; mGluR3
Gene Aliases: 0710001G23Rik; GLUR3; GPRC1C; GRM3; mGlu3; MGLUR3
UniProt ID: (Human) Q14832, (Mouse) Q9QYS2, (Rat) P31422
Entrez Gene ID: (Human) 2913, (Mouse) 108069, (Rat) 24416
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.