Invitrogen
Manufacturer Code:PA536519
Catalog # PIPA536519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 836-886 of human mGluR-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member C; glutamate receptor homolog; glutamate receptor, metabotropic 2; glutamate receptor, metabotropic 3; Metabotropic glutamate receptor 2; Metabotropic glutamate receptor 3; Metabotropic glutamate receptor 3mGluR3GRM3GPRC1C MGLUR3; mGluR2; mGluR3
Gene Aliases: 0710001G23Rik; 4930441L02Rik; GLUR2; GLUR3; GPRC1B; GPRC1C; GRM2; GRM3; mGlu2; mGlu3; MGLUR2; MGLUR3; mGluR7
UniProt ID: (Human) Q14416, (Human) Q14832, (Mouse) Q14BI2, (Rat) P31421, (Mouse) Q9QYS2, (Rat) P31422
Entrez Gene ID: (Human) 2912, (Human) 2913, (Mouse) 108068, (Rat) 24415, (Mouse) 108069, (Rat) 24416
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.