Invitrogen
Manufacturer Code:PA146113
Catalog # PIPA146113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Residues 860-872 of the rat mGlurR2/3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Metabotropic glutamate receptor 2; Metabotropic glutamate receptor 3; mGluR2; MGLUR2 mGlu2 Metabotropic glutamate receptor 2 Metabotropic glutamate receptor 3 GPRC1B GRM2 GRM3 GPRC1C; mGluR3
Gene Aliases: Gprc1b; Gprc1c; Grm2; Grm3; Mglur2; Mglur3
UniProt ID: (Rat) P31421, (Rat) P31422
Entrez Gene ID: (Rat) 24415, (Rat) 24416
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.