Novus Biologicals
Manufacturer Code:NBP267900
Catalog # NBP267900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JJ091-09 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: GLUR2 glutamate metabotropic receptor 2 glutamate receptor metabotropic 2 GPRC1Bmetabotropic glutamate receptor 2 mGlu2 mGluR2 MGLUR2glutamate receptor homolog; glutamate receptor homolog; glutamate receptor, metabotropic 2; Metabotropic glutamate receptor 2; mGluR2
Gene Aliases: GLUR2; GPRC1B; GRM2; mGlu2; MGLUR2
UniProt ID: (Human) Q14416
Entrez Gene ID: (Human) 2912
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.