Proteintech
Manufacturer Code:199561AP150UL
Catalog # 50-173-2622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; GLUR2 GPRC1B GRM2 mGlu2 mGluR2; glutamate receptor homolog; glutamate receptor, metabotropic 2; Metabotropic glutamate receptor 2; mGluR2
Gene Aliases: 4930441L02Rik; GLUR2; GPRC1B; GRM2; mGlu2; MGLUR2; mGluR7
UniProt ID: (Human) Q14416, (Mouse) Q14BI2, (Rat) P31421
Entrez Gene ID: (Human) 2912, (Mouse) 108068, (Rat) 24415
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.